Learn More
Alpaca anti-Rabbit IgG Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 488, Invitrogen™
Alpaca Recombinant Monoclonal Antibody
Marke: Invitrogen SA510323
Weitere Details : Netto-Gewicht : 0.01000kg
Description
Biclonal nanobodies specific for rabbit IgG Fab and Fc. CTK0101: KD = 0.2 nM, CTK0102: KD = 1.2 nM Non Cross-Reactivity includes: Mouse, rat, sheep, goat, and guinea pig serum. Cross-reactivity to: Human and macaque serum Rapid workflow: Simultaneous incubation of rabbit IgG primary antibody and Alpaca anti-rabbit IgG Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek.
Rabbit IgG Nano (VHH) Recombinant Monoclonal antibody specifically detects Rabbit IgG Nano (VHH) in Rabbit samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western BlotSpecifications
Rabbit IgG Nano (VHH) | |
Monoclonal | |
0.5 mg/mL | |
10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7 | |
Alpaca | |
Affinity chromatography | |
RUO | |
Rabbit | |
Liquid |
Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot | |
CTK0101/CTK0102 | |
Alexa Fluor 488 | |
Biclonal; Nanobodies; recombinant VHH | |
Purified rabbit IgG | |
100 μL | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |