Learn More
IgM Monoclonal Antibody (II/41), APC-eFluor™ 780, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
97.00 € - 297.00 €
Spezifikation
Antigen | IgM |
---|---|
Klon | II/41 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15315129
|
Affymetrix eBioscience
47-5790-82 |
100 μg |
297.00 €
100 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen | Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15305129
|
Affymetrix eBioscience
47-5790-80 |
25 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The II/41 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated II/41 can be used as a detection secondary for mouse IgM. Applications Reported: This II/41 antibody has been reported for use in flow cytometric analysis. Applications Tested: This II/41 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.Spezifikation
IgM | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
16019 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
II/41 | |
Flow Cytometry | |
APC-eFluor 780 | |
Rat | |
Mouse | |
Ighm | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Ighm |
For Research Use Only.