Learn More
IKK epsilon Monoclonal Antibody (6B4B5), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515377
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15377 targets IKBKE in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15377 immunogen is purified recombinant fragment of IKBKE (aa1-257) expressed in E. Coli.
IKK epsilon Monoclonal antibody specifically detects IKK epsilon in Human samples. It is validated for ELISA, Western Blot.Spezifikation
IKK epsilon | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q14164 | |
IKBKE | |
Purified recombinant fragment of IKBKE (aa1-257) expressed in E. Coli. | |
RUO | |
9641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
6B4B5 | |
Unconjugated | |
IKBKE | |
AW558201; I-kappa-B kinase epsilon; Ikbke; IKKE; IKK-E; IKKepsilon; IKK-epsilon; IKKI; IKK-i; IKK-related kinase epsilon; inducible I kappa-B kinase; inducible IkappaB kinase; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon; inhibitor of kappaB kinase epsilon; inhibitor of nuclear factor kappa-B kinase subunit epsilon; KIAA0151; RP11-534L20.1 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |