missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13 Monoclonal Antibody (32007), PE, Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA523560
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Flow Cytometry.Spezifikation
IL-13 | |
Monoclonal | |
PE | |
Il13 | |
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
Mouse | |
Protein A/G | |
RUO | |
3596 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
32007 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P35225 | |
Il13 | |
Human IL-13 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |