missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15R alpha Antibody (RM0103-6C38) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Marke: Novus Biologicals NBP1-22466-0.025MG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Mouse samples. It is validated for Western Blot.
Spezifikation
| IL-15R alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 3601 | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot | |
| RM0103-6C38 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| IL15RA | |
| Mouse recombinant protein of the extracellular domain of IL-15RA | |
| 0.025 mg | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-15RA. | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur