Learn More
IL-2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547217
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In sandwich ELISAs, approximately 3% cross-reactivity with recombinant canine IL-2 is observed, approximately 1% cross reactivity with recombinant human IL-2 and recombinant porcine IL-2 is observed, and less than 0.2% cross-reactivity with recombinant mouse IL-2, recombinant equine IL-2, recombinant bovine IL-2, recombinant rat IL-2, and recombinant cotton rat IL-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-2 Polyclonal antibody specifically detects IL-2 in Feline samples. It is validated for ELISA, Immunocytochemistry, Neutralization, Western Blot.Spezifikation
IL-2 | |
Polyclonal | |
Unconjugated | |
IL2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
Goat | |
Antigen affinity chromatography | |
RUO | |
751114 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q07885 | |
IL2 | |
E. coli-derived recombinant feline IL-2 Ala21-Thr154 (Cys146Ser) | |
100 μg | |
Primary | |
Feline | |
Antibody | |
IgG |