Learn More
PeproTech IL-4 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
219.00 € - 3435.00 €
Spezifikation
| Antigen | IL-4 |
|---|---|
| Konzentration | 0.1-1.0 mg/mL |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Biotin |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
17823063
|
PeproTech
500-P94BT-25UG |
25 μg |
219.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
17803093
|
PeproTech
500-P94BT-50UG |
50 μg |
275.00 €
50 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
17863083
|
Invitrogen
500-P94BT-1MG |
2 x 500μg | |||||||
|
30054509
|
PeproTech
500-P94BT-500UG |
500 μg |
1836.00 €
500 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
AA Sequence of recombinant protein: MHGCNDSPLR EIINTLNQVT EKGTPCTEMF VPDVLTATRN TTENELICRA SRVLRKFYFP RDVPPCLKNK SGVLGELRKL CRGVSGLNSL RSCTVNESTL TTLKDFLESL KSILRGKYLQ SCTSMS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-4. Anti-Rat IL-4-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat IL-4 (500-P94) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-4. Western Blot: To detect Rat IL-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Rat IL-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P94BT-1MG will be provided as 2 x 500 μg
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Spezifikation
| IL-4 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Rat | |
| P20096 | |
| 287287 | |
| E.coli-derived Recombinant Rat IL-4. | |
| Antigen affinity chromatography | |
| Il4 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
| Il4 | |
| Primary | |
| -20°C |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.