Learn More
IL-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 39 publications
Marke: Novus Biologicals NB600-1131SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-6 Polyclonal specifically detects IL-6 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Spezifikation
| IL-6 | |
| Polyclonal | |
| RUO |
| Western Blot, ELISA, Immunohistochemistry | |
| Rabbit | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |