Learn More
IL-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 39 publications
232.00 € - 532.00 €
Spezifikation
| Antigen | IL-6 |
|---|---|
| Anwendungen | Western Blot, ELISA, Immunohistochemistry |
| Klassifikation | Polyclonal |
| Wirtsspezies | Rabbit |
| Regulatorischer Status | RUO |
| Produktcode | Marke | dimQuantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | dimQuantity | Preis | Menge & Verfügbarkeit | |||||
|
18728083
|
Novus Biologicals
NB600-1131SS |
232.00 €
0.04 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | ||||||
|
18790664
|
Novus Biologicals
NB600-1131 |
532.00 €
0.20 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | ||||||
Beschreibung
IL-6 Polyclonal specifically detects IL-6 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.Spezifikation
| IL-6 | |
| Polyclonal | |
| RUO |
| Western Blot, ELISA, Immunohistochemistry | |
| Rabbit | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |