Learn More
IL1RL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538013
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
A recommended positive control is human small intestine tissue lysate. At least three isoforms of IL-1RL2 are known to exist; this antibody will detect all three isoforms. IL-1RL2 antibody is not expected to cross-react with IL-1R or IL-1RL1.
IL1RL2 Polyclonal antibody specifically detects IL1RL2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IL1RL2 | |
Polyclonal | |
Unconjugated | |
IL1RL2 | |
AI481289; AW551444; IL-1 receptor-related protein 2; IL1RL2; IL1RRP2; IL-1Rrp2; IL1R-rp2; IL-36 receptor; IL-36R; interleukin 1 receptor like 2; interleukin 1 receptor-like 2; interleukin-1 receptor related protein 2; interleukin-1 receptor-like 2; Interleukin-1 receptor-related protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107527, 171106, 8808 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q62929, Q9ERS7, Q9HB29 | |
IL1RL2 | |
16 amino acid peptide near the center of human IL-1RL2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |