missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL36RN Mouse anti-Human, Clone: 5F11G6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60298-1-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.Spezifikation
IL36RN | |
Monoclonal | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UBH0 | |
IL36RN | |
IL36RN Fusion Protein Ag3780 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
5F11G6 | |
Unconjugated | |
IL36RN | |
FIL1, FIL1 delta, FIL1(DELTA), FIL1D, IL 1 delta, IL 1 related protein 3, IL 1F5, IL 1HY1, IL 1L1, IL 1ra homolog 1, IL 1RP3, IL1F5, IL1HY1, IL1L1, IL1RP3, Interleukin 1 delta, Interleukin 1 family member 5, Interleukin 1 HY1, Interleukin 1 like protein 1 | |
Mouse | |
Protein A | |
RUO | |
26525 | |
-20°C | |
Liquid |