Learn More
Imidazo-[1,2-a]-pyridin-6-carbonsäure Hydrazid, 97 %, Thermo Scientific™
Click to view available options
Menge:
250mg
1g
5g
886361-97-3
C8H8N4O
176.179
MFCD07021499
ZMQNWDRVASCREF-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carbohydrazide, imidazo 1,2-a pyridine-6-carboxylic acid hydrazide, imidazoapyridinecarbohydrazide, 4-hydroimidazo 1,2-a pyridine-6-carbohydrazide
16413557
Imidazo[1,2-a]Pyridin-6-Carbohydrazid
C1=CC2=NC=CN2C=C1C(=O)NN
886361-97-3
C8H8N4O
176.179
MFCD07021499
ZMQNWDRVASCREF-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carbohydrazide, imidazo 1,2-a pyridine-6-carboxylic acid hydrazide, imidazoapyridinecarbohydrazide, 4-hydroimidazo 1,2-a pyridine-6-carbohydrazide
16413557
Imidazo[1,2-a]Pyridin-6-Carbohydrazid
C1=CC2=NC=CN2C=C1C(=O)NN
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemischer Name oder Material Imidazo[1,2-a]pyridine-6-carboxylic acid hydrazide
Schmelzpunkt 213°C to 215°C
Menge 250mg
Formelmasse 176.18
Reinheit (%) 97%