Learn More
STEMCELL Technologies ImmunoCult™ Human Th1 Differentiation Supplement
Artikelnummer. 17197641
Encompass
Change view
Click to view available options
Menge:
1 ml
Artikelnummer. Menge
17197641 1 ml
1 options
Artikelnummer. 17197641
Marke: STEMCELL Technologies 10973
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Culture supplement for the differentiation of human naïve CD4+ T cells into Th1 cells
  • ImmunoCult™ Human Th1 Differentiation Supplement contains a combination of human cytokines and a blocking antibody formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into Th1 cells
  • ImmunoCult™ Human Th1 Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981) and ImmunoCult™ Human CD3/CD28 T Cell Activator (Catalog #10971)
  • This product is recommended for: Research into the regulation of human naïve T cell differentiation and Th1 function
  • Development of procedures to expand and polarize naïve T cells toward a Th1 subset
Spezifikation
Zelltyp T Cells, T Cells, CD4+
Zur Verwendung mit (Anwendung) Cell Culture, Differentiation, Expansion
Menge 1 ml