Learn More
Importin 8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554881
Detalles adicionales : Netto-Gewicht : 0.02000kg
Descripción
Immunogen sequence: SNNGRGEDEE EEDDDWDEEV LEETALEGFS TPLDLDNSVD EYQFFTQALI TVQSRDAAWY QLLMAPLSED QRTALQEVYT LAEHRRTVAE AKKKIEQQGG FTFENKGVLS AFNFGTVPSN N Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 44%.
Importin 8 is a 1037 amino acid nuclear protein with a RAN binding site. It belongs to RAN-binding protein super family and hence shares an N-terminal sequence motif with importin-beta that appears to account for RAN GTP binding. It binds to the nuclear pore complex (NPC) and is well known as a transport carrier that mediates nuclear import of ribosomal proteins with a classical nuclear localization signal. Importin 8 works by binding to the nuclear pore complex and, along with RANGTP and RANBP1, inhibits the GAP stimulation of the RAN GTPase. Along with transportin it also helps in the nuclear import of SRP19.Especificaciones
Importin 8 | |
Polyclonal | |
Unconjugated | |
IPO8 | |
6230418K12Rik; Abcc10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; C130009K11Rik; FLJ26580; imp8; importin 8; importin-8; IPO8; MRP7; Om1; OM-1; oocyte maturation 1; RAN binding protein 8; ran-binding protein 8; RANBP8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15397 | |
IPO8 | |
Recombinant protein corresponding to Human Importin 8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only