missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B HA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534975
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: virus infected medium_2 (Taiwan B70555). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.Spezifikation
Influenza B HA | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; Influenza B; influenza b hemagglutinin; Influenza HA; influenza hemagglutinin | |
Recombinant fragment contains a sequence corresponding to a region within amino acids and of Influenza B Virus Hemagglutinin (HA) | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |