missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B NA Monoclonal Antibody (B14), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110824
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-10824 detects Influenza B Neuraminidase. MA1-10824 has been successfully used in ELISA and inhibition applications. The MA1-10824 immunogen is Influenza B HK/73.
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.Spezifikation
Influenza B NA | |
Monoclonal | |
0.88 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Inhibition Assays | |
B14 | |
Unconjugated | |
Influenza NA | |
Influenza B HK/73. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |