missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INH6, MedChemExpress
Beschreibung
INH6 is a potent Nek2/Hec1 inhibitor
Spezifikation
Spezifikation
| Chemischer Name oder Material | INH6 |
| Physikalische Form | Solid |
| CAS | 1001753-24-7 |
| Farbe | White |
| Menge | 100 mg |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C19H18N2OS |
| Zur Verwendung mit (Anwendung) | Cancer-programmed cell death |
| Löslichkeitsinformationen | DMSO : 50 mg/mL (155.08 mM; Need ultrasonic) |
| SMILES | O=C(NC1=NC(C2=C(C)C=C(C)C=C2C)=CS1)C3=CC=CC=C3 |
| Mehr anzeigen |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?