missing translation for 'onlineSavingsMsg'
Learn More

INH6, MedChemExpress

Product Code. p-200096140
Encompass_Preferred
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
Product Code. Menge
16449636 50 mg
16423078 10 mg
16404376 100 mg
3 options
This item is not returnable. View return policy

Product Code. 16449636

Brand: Medchem Express HY100541

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress INH6 is a potent Nek2/Hec1 inhibitor; inhibits the growth of HeLa cells with an IC50 of 2.4 μM.

INH6 is a potent Nek2/Hec1 inhibitor

Specifications

Chemischer Name oder Material INH6
Physikalische Form Solid
CAS 1001753-24-7
Farbe White
Menge 50 mg
Hinweise zur Reinheitsqualität Research
Summenformel C19H18N2OS
Zur Verwendung mit (Anwendung) Cancer-programmed cell death
Löslichkeitsinformationen DMSO : 50 mg/mL (155.08 mM; Need ultrasonic)
SMILES O=C(NC1=NC(C2=C(C)C=C(C)C=C2C)=CS1)C3=CC=CC=C3
Molekulargewicht (g/mol) 322.42
Formelmasse 322.42
Reinheit (%) 99.9%
Güte Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.