Learn More
INO80C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561869
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GYGASKKKKA SASSFAQGIS MEAMSENKMV PSEFSTGPVE KAAKPLPFKD PNFVHSGHGG AVAGKKNRT Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
INO80C is a proposed core component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair.Spezifikation
INO80C | |
Polyclonal | |
Unconjugated | |
INO80C | |
C18orf37; hIes6; IES6; IES6 homolog; INO80 complex subunit C; INO80C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
125476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PI98 | |
INO80C | |
Recombinant protein corresponding to Human INO80C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only