Learn More
Eurofins DiscoverX Insulin Receptor Protein, Active
Beschreibung
The insulin receptor kinase (IRK) is composed of two extracellular a-subunits (135 kDa), which contain the insulin-binding site, and two intracellular b-subunits (95 kDa), containing the protein tyrosine kinase domain. After insulin binding, the insulin-IRK complex internalizes into muscle and fat cells. The IRK substrates of IRS-1 and IRS-2 form complexes, through Src homology region 2 (SH2) domains, with docking molecules such as phosphoinositde-3-kinase (PI3K). The recruitment of PI3K in turn activates phosphoinositide-dependent kinases, which serve in the activation of protein kinase B (Akt) and atypical isoforms of protein kinase C (PKC). These activated kinases then phosphorylate downstream effectors, ultimately promoting the translocation of insulin-sensitive glucose transporter subtype 4 (GLUT-4) to the plasma membrane, to operate in the coordination of glucose metabolism.
GenBank NM_000208
Spezifikation
Spezifikation
| Zusammensetzung | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekulargewicht | 36.8 kDa |
| Menge | 10 μg |
| Spezies | Human |
| Rekombinant | Recombinant |
| Proteinmarkierung | N-terminal 6His |
| Expressionssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Truncated |
| Reinheits- oder Qualitätsgrad | ≥73% |
| Protein | Insulin Receptor, active |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.