Learn More
Integrin Beta 3 Mouse anti-Human, Unconjugated, Clone: 3D8E5, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66952-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Integrin Beta 3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Integrin Beta 3. This antibody recognizes human antigen. The Integrin Beta 3 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| Integrin Beta 3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry | |
| 3D8E5 | |
| Unconjugated | |
| 150 μL | |
| 3690 | |
| IgG2b |