missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25277-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Protein kinases comprise a large group of encoded factors that regulate cellular processes by catalyzing the transfer of a phosphate group to a hydroxyl acceptor in serine, threonine or tyrosine residues. Myotonic dystrophy kinaserelated Cdc42-binding (DMPK-like) kinases-alpha and beta (MRCK-alpha, beta) contain a cysteine-rich motif and a putative pleckstrin homology domain. MRCKs can phosphorylate nonmuscle Myosin light chain and influences Actin-Myosin contractility. MRCK-alpha can phosphorylate and activate LIM kinases downstream of Cdc42, which leads to inactivation of ADF/Cofilin and to Actin cytoskeletal reorganization. MRCK-alpha can also influence neurite outgrowth promoted by Cdc42 and Rac.Spezifikation
Integrin beta-4 | |
Polyclonal | |
Unconjugated | |
ITGB4 | |
CD104, GP150, Integrin beta 4, integrin, beta 4, ITGB4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192897, 25724, 3691 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2A863, P16144, Q64632 | |
ITGB4 | |
Integrin beta-4 Fusion Protein Ag16460 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |