missing translation for 'onlineSavingsMsg'
Learn More

IP6K1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marke:  Novus Biologicals NBP2-99495-50ul

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 18630486

  • 216.00 € / 50 Mikroliter

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

IP6K1 Polyclonal antibody specifically detects IP6K1 in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

IP6K1
Polyclonal
Immunoprecipitation 1-5uL/mg of lysate
ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IP6K1.
50 μg
Neuroscience, Neurotransmission, Signal Transduction
9807
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunoprecipitation
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.