missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Islet 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15661-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.Spezifikation
Islet 1 | |
Polyclonal | |
Unconjugated | |
ISL1 | |
Isl 1, ISL LIM homeobox 1, ISL1, Islet 1, ISLET1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16392, 3670, 64444 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61371, P61372, P61374 | |
Isl1 | |
Islet 1 Fusion Protein Ag8088 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |