Learn More
ITGA4 Monoclonal Antibody (5C4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517102
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17102 targets ITGA4 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17102 immunogen is purified recombinant fragment of human ITGA4 expressed in E. Coli. MA5-17102 detects ITGA4 which has a predicted molecular weight of approximately 115kDa.
The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.Spezifikation
ITGA4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P13612 | |
ITGA4 | |
Purified recombinant fragment of human ITGA4 expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
5C4 | |
Unconjugated | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Mouse | |
Protein G | |
RUO | |
3676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |