missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGB1 Monoclonal Antibody (HM beta 1-1), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Marke: Invitrogen MA516708
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
ITGB1 Monoclonal antibody specifically detects ITGB1 in Mouse, Rat samples. It is validated for Flow Cytometry.Spezifikation
ITGB1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P09055, P49134 | |
ITGB1 | |
Purified mouse VLA-4 antigen | |
500 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry | |
HM beta 1-1 | |
FITC | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Armenian Hamster | |
Protein G | |
RUO | |
16412, 24511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |