Learn More
ITL BioMedical ITL Corporation Sicherheits-Subkultur-Einheit
Artikelnummer. 15235636
Encompass_Preferred
Change view
Click to view available options
Produkttyp:
Sicherheitseinheit für Subkulturen
Packungsgröße:
60 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Produkttyp unitSize
15235636 Sicherheitseinheit für Subkulturen 60 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15235636 Lieferant ITL BioMedical Lieferanten-Nr. A100720
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Needleless safety device for subculturing blood culture bottles.
Used to transfer sample from positive blood culture bottles for further testing, bringing a new level of safety to microbiology labs.
  • Easy to use and versatile
  • Two procedure options to obtain samples for further testing: drop sample method and syringe method
  • Neither method requires needle which reduces the risk of potential needle stick injury
  • Does not contain latex
  • Compatible fit with various culture bottles
  • Limits exposure to gas or culture discharge
  • Single use only
Spezifikation
Produkttyp Sicherheitseinheit für Subkulturen