Learn More
ITPKA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559185
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DGSCSTDFKT TRSREQVLRV FEEFVQGDEE VLRRYLNRLQ QIRDTLEVSE FFRRHEV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro.Spezifikation
ITPKA | |
Polyclonal | |
Unconjugated | |
ITPKA | |
EC 2.7.1.127; inositol 1,4,5-triphosphate 3-kinase; inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3 kinase A; IP3 3-kinase A; IP3-3KA; IP3K A; IP3KA; IP3-kinase A; ITPKA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228550, 3706 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23677, Q8R071 | |
ITPKA | |
Recombinant protein corresponding to Human ITPKA | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only