missing translation for 'onlineSavingsMsg'
Learn More

Ivabradine metabolite N-Demethyl Ivabradine hydrochloride, MedChemExpress

Artikelnummer. 16468057
Encompass_Preferred
Change view
Click to view available options
Menge:
5 mg
Unit Size:
5 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16468057 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468057 Supplier Medchem Express Supplier No. HY12778

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress N-Demethyl Ivabradine Hcl is a metabolite of Ivabradine, which is a specific inhibitor of the funny channel.

N-Demethyl Ivabradine Hcl is a metabolite of Ivabradine, which is a specific inhibitor of the funny channel.

Specifications

Chemischer Name oder Material Ivabradine metabolite N-Demethyl Ivabradine hydrochloride
CAS 1246638-08-3
Farbe White
Hinweise zur Reinheitsqualität Research
Summenformel C26H35ClN2O5
Menge 5 mg
Synonym N-Demethyl ivabradine hydrochloride
SMILES O=C1N(CCCNC[C@@H]2C(C=C(OC)C(OC)=C3)=C3C2)CCC4=CC(OC)=C(OC)C=C4C1.[H]Cl
Molekulargewicht (g/mol) 491.02
Formelmasse 491.02
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.