Learn More
IWP L6, MedChemExpress
Artikelnummer. 16486848
Encompass_Preferred
Change view
Click to view available options
Menge:
10 mg
50 mg
Packungsgröße:
10 Milligramm
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16486848 10 mg 10 Milligramm
16472917 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16486848 Lieferant Medchem Express Lieferanten-Nr. HY15825
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress IWP L6 (Porcn Inhibitor III) is a Porcn inhibitor with an EC50 of 0.5 nM.
IWP L6 (Porcn Inhibitor III) is a Porcn inhibitor with an EC50 of 0.5 nM.
Spezifikation
Chemischer Name oder Material IWP L6
CAS 1427782-89-5
Farbe White
Hinweise zur Reinheitsqualität Research
Summenformel C25H20N4O2S2
Menge 10 mg
Synonym Porcn Inhibitor III
Löslichkeitsinformationen DMSO : 1.43 mg/mL (3.03 mM; Need ultrasonic)
SMILES O=C(NC1=NC=C(C2=CC=CC=C2)C=C1)CSC3=NC(CCS4)=C4C(N3C5=CC=CC=C5)=O
Molekulargewicht (g/mol) 472.58
Formelmasse 472.58
Reinheit (%) 99.02%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease