missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JAB1 Recombinant Rabbit Monoclonal Antibody (23GB3490), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA552610
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the Signalosome, which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
Spezifikation
| JAB1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cops5 | |
| 38 kDa Mov34 homolog; AI303502; COP9 (constitutive photomorphogenic) homolog, subunit 5; COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana); COP9 complex S5; COP9 constitutive photomorphogenic homolog subunit 5; COP9 signalosome complex subunit 5; COP9 signalosome subunit 5; Cops5; CSN5; JAB1; Jun activation domain-binding protein 1; Jun coactivator; Kic2; Kip1 C-terminus-interacting protein 2; MGC3149; Mov34; MOV-34; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10987, 26754, 312916 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB3490 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35864, Q92905 | |
| Cops5 | |
| A synthetic peptide of human JAB1 (10-80AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur