Learn More
Jagged1 Mouse anti-Human, Unconjugated, Clone: 2A7D11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66890-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Jagged1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Jagged1. This antibody recognizes human antigen. The Jagged1 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| Jagged1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Flow Cytometry | |
| 2A7D11 | |
| Unconjugated | |
| 20 μL | |
| 182 | |
| IgA |