Learn More
KATNA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557571
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RVHRSSAQNV HNDRGKAVRC REKKEQNKGR EEKNKSPAAV TEPETNKFDS TGYDKDLVEA LERDIISQNP NVRWDD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.Spezifikation
KATNA1 | |
Polyclonal | |
Unconjugated | |
KATNA1 | |
HAMAP-Rule:MF_03023}; hypothetical protein LOC553631; katanin catalytic subunit A1; katanin p60 (ATPase containing) subunit A 1; katanin p60 (ATPase-containing) subunit A 1; katanin p60 (ATPase-containing) subunit A1; katanin p60 ATPase-containing subunit A1; katanin p60 ATPase-containing subunit A1 {ECO:0000255; Katanin p60 ATPase-containing subunit A1-like protein; katanin p60 subunit A 1; katanin p60 subunit A1; katanin p60 subunit A1 {ECO:0000255; KATNA1; lipotransin; microtubule severing protein; p60 katanin; p60 katanin {ECO:0000255; si:dkey-15j16.1; zgc:110580 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11104 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75449 | |
KATNA1 | |
Recombinant protein corresponding to Human KATNA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only