Learn More
KCNK18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560810
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ILSTSYNRFR KFPFFTRPLL SKWCPKSLFK KKPDPKPADE AVPQIIISAE ELPGPKLGTC PSR Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 48%.
KCNK18 Polyclonal antibody specifically detects KCNK18 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KCNK18 | |
Polyclonal | |
Unconjugated | |
KCNK18 | |
2P K ion channel TRESK; Gm781; K2p18.1; Kcnk18; MGR13; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; potassium channel, two pore domain subfamily K, member 18; potassium two pore domain channel subfamily K member 18; tandem-pore K+ (K(2P)) channel; Tresk; TRESK2; Tresk-2; TRIK; TWIK-related individual K+ channel; TWIK-related individual potassium channel; TWIK-related spinal cord K+ channel; TWIK-related spinal cord potassium channel; Two-pore-domain potassium channel TRESK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
338567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z418 | |
KCNK18 | |
Recombinant protein corresponding to Human KCNK18 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only