Learn More
KHDRBS3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541821
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EQSYDSYDNS YSTPAQSGAD YYDYGHGLSE ETYDSYGQEE WTNSRHKAPS Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 100%
KHDRBS3 Polyclonal antibody specifically detects KHDRBS3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KHDRBS3 | |
Polyclonal | |
Unconjugated | |
Khdrbs3 | |
Etle; etoile; etoile, Sam68-like protein SLM-2; KH domain containing, RNA binding, signal transduction associated 3; KH domain-containing, RNA-binding, signal transduction-associated protein 3; KH RNA binding domain containing, signal transduction associated 3; KHDRBS3; RNA-binding protein Etoile; RNA-binding protein T-Star; rSLM-2; SALP; Sam68-like mammalian protein 2; Sam68-like phosphotyrosine protein; Sam68-like phosphotyrosine protein, T-STAR; SLM2; SLM-2; TSTAR; T-STAR | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10656 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75525 | |
Khdrbs3 | |
Synthetic peptide directed towards the C-terminal of human KHDRBS3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |