Learn More
KHDRBS3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551392
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GEGKDEEKYI DVVINKNMKL GQKVLIPVKQ FPKFNFVGKL LGPRGNSLKR LQEETLTKMS ILGKGSMRDK AKEEELRKSG EAKYFHLNDD LHVLIEVFAP PAEAYARM Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
As a RNA-binding protein, KHDRBS3 plays a role in the regulation of alternative splicing and influences mRNA splice site selection and exon inclusion. KHDRBS3 may play a role as a negative regulator of cell growth. It inhibits cell proliferation and involved in splice site selection of vascular endothelial growth factor. It induces an increased concentration-dependent incorporation of exon in CD44 pre-mRNA by direct binding to purine-rich exonic enhancer. RNA-binding abilities are down-regulated by tyrosine kinase PTK6. It also involved in post-transcriptional regulation of HIV-1 gene expression.Spezifikation
KHDRBS3 | |
Polyclonal | |
Unconjugated | |
Khdrbs3 | |
Etle; etoile; etoile, Sam68-like protein SLM-2; KH domain containing, RNA binding, signal transduction associated 3; KH domain-containing, RNA-binding, signal transduction-associated protein 3; KH RNA binding domain containing, signal transduction associated 3; KHDRBS3; RNA-binding protein Etoile; RNA-binding protein T-Star; rSLM-2; SALP; Sam68-like mammalian protein 2; Sam68-like phosphotyrosine protein; Sam68-like phosphotyrosine protein, T-STAR; SLM2; SLM-2; TSTAR; T-STAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10656, 13992, 64015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75525, Q9JLP1, Q9R226 | |
Khdrbs3 | |
Recombinant protein corresponding to Human KHDRBS3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only