missing translation for 'onlineSavingsMsg'
Learn More

Ki-67 Monoclonal Antibody (SolA15), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marke:  Affymetrix eBioscience 53-5698-82

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16320994

  • 374.00 € / 1 Stück
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Description: The monoclonal antibody SolA15 recognizes mouse and rat Ki-67, a 300 kDa nuclear protein. Ki-67 is present during all active phases of the cell cycle (G1, S, G2, and mitosis), but is absent from resting cells (G0). Ki-67 is detected within the nucleus during interphase but redistributes to the chromosomes during mitosis. Ki-67 is used as a marker for determining the growth fraction of a given population of cells. In studies of tumor cells, the "Ki-67 labeling index" refers to the number of Ki-67 positive cells within the population and this is used to predict outcome of particular cancer types. Ki-67 has been shown to interact with the DNA-bound protein chromobox protein homolog 3 (CBX3) (heterochromatin). The SolA15 antibody also recognizes human, non-human primate and canine Ki-67. Applications Reported: This SolA15 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This SolA15 antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Refer to Best Protocols for Staining Protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

Ki-67 is a nuclear protein that is expressed during various stages in the cell cycle, particularly during late G1, S, G2, and M phases. The protein has a forkhead associated domain (FHA) through which it associates with euchromatin at the perichromosomal layer, the centromeric heterochromatin, and the nucleolus. Ki-67 is shown to have a cell cycle dependent topographical distribution with perinucleolar expression at G1, expression in the nuclear matrix at G2, and expression on the chromosomes during M phase. Ki-67 is commonly used as a proliferation marker because it is not detected in G0 cells, but increases steadily from G1 through mitosis. Ki-67 antibodies are useful in establishing the cell growing fraction in neoplasms. In neoplastic tissues, the prognostic value is comparable to the tritiated thymidine-labelling index. The correlation between low Ki-67 index and histologically low-grade tumors is strong. Ki-67 is routinely used as a neuronal marker of cell cycling and proliferation.
Spezifikation

Spezifikation

Ki-67
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide, pH 7.2
E9PVX6, P46013
Mki67
Affinity chromatography
RUO
100686578, 102135895, 17345, 291234, 4288
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
SolA15
Alexa Fluor 488
Mki67
antigen identified by monoclonal antibody Ki 67, antigen identified by monoclonal antibody Ki-67, Antigen identified by monoclonal antibody Ki-67 homolog, Antigen KI-67, Antigen KI-67 homolog, antigen KI-67, proliferation marker protein Ki-67, antigen KI-67-like, cb31, D630048A14Rik, I79_022666, Ki67, Ki-67, KIA, LOW QUALITY PROTEIN: proliferation marker protein Ki-67, marker of proliferation Ki-67, MIB-, MIB-1, Mki67, PPP1R105, Proliferation marker protein Ki-67, proliferation-related Ki-67 antigen, protein phosphatase 1, regulatory subunit 105, RP11-380J17.2, sb:cb31, si:ch211-250b22.7, unnamed protein product, wu:fa11g09, wu:fb57a07, wu:fi14e05
Rat
100 μg
Primary
Canine, Cynomolgus Monkey, Human, Mouse, Non-human Primate, Rat
Antibody
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote