Learn More
KIAA1033 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561064
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KVLNKVITVY AALCCEIKKL KYEAETKFYN GLLFYGEGAT DASMVEGDCQ IQMGRFISFL QELSCFVTRC YEVVMNVVHQ LAALYISNKI APKII Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 91%.
This gene encodes a component of the WASH complex, which functions in the intracellular transport of endosomes. Mutations in this gene have been detected in individuals with autosomal recessive cognitive disability. Alternative splicing results in multiple transcript variants.Spezifikation
KIAA1033 | |
Polyclonal | |
Unconjugated | |
WASHC4 | |
A230046K03Rik; AA589518; Kiaa1033; mKIAA1033; MRT43; RIKEN cDNA A230046K03 gene; strumpellin and WASH-interacting protein; SWIP; WASH complex subunit 4; WASH complex subunit 7; WASH complex subunit SWIP; WASHC4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2M389 | |
WASHC4 | |
Recombinant protein corresponding to Human KIAA1033 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only