Learn More
KLHDC7A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563330
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CATYRTPYPD AFQCAVVDNL IYCVGRRSTL CFLADSVSPR FVPKELRSFP APQGTLLPTV LTLPTPDLPQ TR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.
KLHDC7A Polyclonal antibody specifically detects KLHDC7A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
KLHDC7A | |
Polyclonal | |
Unconjugated | |
KLHDC7A | |
kelch domain containing 7A; kelch domain-containing protein 7A; KLHDC7A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127707 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VTJ3 | |
KLHDC7A | |
Recombinant protein corresponding to Human KLHDC7A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only