Learn More
KLHL11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562944
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NSDDIDKQYR KEAYRYCAER KRWMLLPPMP QPRCRATACH VRIPYRYLHG TQRYPMPQNL MWQKDRIRQM QEIHRHALNM RRVPSSQIEC Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Kelch-like protein 11, Subcellular location Secreted.Spezifikation
KLHL11 | |
Polyclonal | |
Unconjugated | |
KLHL11 | |
BC011167; FLJ10572; kelch like family member 11; kelch-like 11; kelch-like family member 11; kelch-like protein 11; KLHL11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NVR0 | |
KLHL11 | |
Recombinant protein corresponding to Human KLHL11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only