missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRT14 (Human) Recombinant Protein
Human KRT14 full-length ORF ( AAH02690, 1 a.a. - 472 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00003861-P01.25ug
Informations supplémentaires : Netto-Gewicht : 0.00010kg
Description
- Sequence: MTTCSRQFTSSSSMKGSCGIGGGIGGGSSRISSVLAGGSCRAPSTYGGGLSVSSSRFSSGGAYGLGGGYGGGFSSSSSSFGSGFGGGYGGGLGTGLGGGFGGGFAGGDGLLVGSEKVTMQNLNDRLASYLDKVRALEEANADLEVKIRDWYQRQRPAEIKDYSPYFKTIEDLRNKILTATVDNANVLLQIDNARLAADDFRTKYETELNLRMSVEADINGLRRVLDELTLARADLEMQIESLKEELAYLKKNHEEEMNALRGQVGGDVNVEMDAAPGVDLSRILNEMRDQYEKMAEKNRKDAEEWFFTKTEELNREVATNSELVQSGKSEISELRRTMQNLEIELQSQLSMKASLENSLEETKGRYCMQLAQIQEMIGSVEEQLAQLRCEMEQQNQEYKILLDVKTRLEQEIATYRRLLEGEDAHLSSSQFSSGSQSSRDVTSSSRQIRTKVMDVHDGKVVSTHEQVLRTKN
Spécification
AAH02690 | |
keratin 14 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRT14 | |
GST |
3861 | |
Wheat germ expression system | |
25 μg | |
CK14, EBS3, EBS4, K14, NFJ | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |