Learn More
KRT74 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561778
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ISVISSSSYS YHHPSSAGVD LGASAVAGSS GSTQSGQTKT TEARGGDLKD TQGKSTPASI PARKATR Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 36%.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This protein belongs to a family of keratins that are specifically expressed in the inner root sheath of hair follicles.Spezifikation
KRT74 | |
Polyclonal | |
Unconjugated | |
KRT74 | |
ADWH; CK-74; cytokeratin-74; HTSS2; HYPT3; K5C; K6IRS4; K74; KB37; keratin 5c; keratin 6 irs4; keratin 74; keratin 74, type II; keratin, type II cytoskeletal 74; Keratin-5c; keratin-74; KRT5C; KRT6IRS4; KRT74; Type II inner root sheath-specific keratin-K6irs4; type-II keratin Kb37 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121391 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTS7 | |
KRT74 | |
Recombinant protein corresponding to Human KRT74 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only