Learn More
STEMCELL Technologies L-Glutamine
Cell culture supplement
Marke: STEMCELL Technologies 07100
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
- L-Glutamine (200 mM) is recommended for use as a cell culture supplement and is required for the preparation of MesenCult™-ACF Plus Medium (Catalog #05445)
Spezifikation
Mesenchymal Stem and Progenitor Cells, Neurons, Other | |
Cell Culture | |
100 mL |