Learn More
L3MBTL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540325
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EYDQWVDCES PDIYPVGWCE LTGYQLQPPV AAEPATPLKA KEATKKKKKQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 80%; Rabbit: 100%; Rat: 86%
L3MBTL2 Polyclonal antibody specifically detects L3MBTL2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
L3MBTL2 | |
Polyclonal | |
Unconjugated | |
L3MBTL2 | |
4732493N06Rik; H-l(3)mbt-l; H-l(3)mbt-like protein 2; l(3)mbt-like 2; l(3)mbt-like 2 (Drosophila); l(3)mbt-like 2 protein; L(3)mbt-like protein 2; L3MBT; L3MBTL histone methyl-lysine binding protein 2; L3MBTL2; L3MBTL2 polycomb repressive complex 1 subunit; L3MBTL2, polycomb repressive complex 1 subunit; lethal(3)malignant brain tumor-like 2 protein; lethal(3)malignant brain tumor-like protein 2; lmbt-like 2 (Drosophila) isoform 1; lmbt-like 2 (Drosophila) isoform 2; M4mbt | |
Rabbit | |
Protein A | |
RUO | |
83746 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q969R5 | |
L3MBTL2 | |
Synthetic peptide directed towards the C-terminal of human L3MBTL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |