Learn More
Epredia™ Lab Vision™ ALK (Anaplastic Lymphoma Kinase)/p80 Rabbit Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ ALK (Anaplastic Lymphoma Kinase)/p80, Rabbit Monoclonal Antibody.
203.00 € - 772.00 €
Spezifikation
| Antigen | ALK (Anaplastic Lymphoma Kinase)/p80 |
|---|---|
| Klon | SP8 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Form | Preis | Menge & Verfügbarkeit | ||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Form | Preis | Menge & Verfügbarkeit | ||||
|
12613717
|
Epredia™
RM-9108-S0 |
100 μL | Concentrated | ||||||
|
12623717
|
Epredia™
RM-9108-S1 |
500 μL | Concentrated | ||||||
|
12683707
|
Epredia™
RM-9108-S |
1 mL | Concentrated | ||||||
|
12603717
|
Epredia™
RM-9108-R7 |
7 mL | Prediluted | ||||||
|
12673707
|
Epredia™
RM-9108-RQ |
12 mL | Ready-to-Use | ||||||
Beschreibung
Recognizes a human p80 protein, identified as a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in 30-50% of CD30+ large cell lymphomas (1). This rabbit monoclonal antibody can be used to detect p80 in these lymphomas and may also be used to detect a recently described subtype of large B cell lymphoma, which expresses the full-length ALK protein (2).Host Species: Rabbit
Clone: SP8
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript
Positive Control: Anaplastic lymphoma
Cellular Localization: Cytoplasmic + nuclear Clone SP8 is excellent for staining of formalin/paraffin with no special pretreatment
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spezifikation
| ALK (Anaplastic Lymphoma Kinase)/p80 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Recombinant protein corresponding to a region, which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript |
| SP8 | |
| Monoclonal | |
| Rabbit | |
| Cancer and Tumor Biology | |
| Primary |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.