Learn More
Collagen II Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.
Marke: Epredia MS-235-R7
Beschreibung
Host Species: Mouse
Clone: 2B1.5
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.
Epitope: Carboxyl-terminal one quarter of the type II collagen. The epitope for 2B1.5 MAb is not susceptible to digestion of lathyritic type II collagen with recombinant stromelysin
Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Positive Control: Cartilage in lung or fetus
Cellular Localization: Cartilage matrix
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Spezifikation
| Collagen II Ab-2 | |
| Monoclonal | |
| Prediluted | |
| Mouse | |
| 7 mL | |
| Extracellular Matrix | |
| Bovine, Chicken, Human, Mouse, Rat | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2B1.5 | |
| Unconjugated | |
| A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
| RUO | |
| Primary | |
| Prediluted |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.