missing translation for 'onlineSavingsMsg'
Learn More

Interferon-α (II)/IFN-α (II) Mouse Monoclonal Antibody, Epredia™

Epredia™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody provides accurate, reproducible results.

Marke:  Epredia MS-574-P1

Weitere Details : Netto-Gewicht : 1.00000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 12601657

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

The IFN-α family consists of 24 or more genes or pseudo-genes. IFN-αII) is one of the two distinct families (I and II) of human IFN-α. The α interferons are mainly produced by lymphocytes, monocytes, macrophages, and cell lines such as Namalwa and KG1 following induction by viruses, nucleic acids, and glucocorticoid hormones. They are involved in virus resistance on target cells, inhibition of cell proliferation, induction of cytokines and regulation of expression of MHC class I antigens.

Host Species: Mouse

Clone: N27

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 43-53

Immunogen: Purified recombinant IFN-α 2c

Molecular Weight: 16-27kDa

Positive Control: Stimulated human peripheral blood mononuclear cells

Recommended for:

  • Blocking study

Note:

USA: RUO; Int'l: RUO

Spezifikation

Spezifikation

Interferon-alpha (II)/IFN-alpha (II)
N27
Unconjugated
Purified recombinant IFN-alpha 2c
RUO
Primary
Concentrated
Monoclonal
200μg/mL
Mouse
500 μL
Cytokines
Human
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote