Learn More
Kappa Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
110.16 € - 347.00 €
Spezifikation
| Antigen | Kappa Light Chain Ab-1 |
|---|---|
| Klon | L1C1 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Wirtsspezies | Mouse |
| Produktcode | Marke | Menge | Konjugat | Form | Konzentration | Preis | Menge & Verfügbarkeit | ||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Konjugat | Form | Konzentration | Preis | Menge & Verfügbarkeit | ||
|
12689796
|
Epredia
MS-143-B0 |
100 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12699796
|
Epredia
MS-143-B1 |
500 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12619806
|
Epredia
MS-143-P0 |
100 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12639806
|
Epredia
MS-143-PABX |
200 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12385743
|
Epredia
MS-143-P1 |
500 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12609806
|
Epredia
MS-143-P |
1 mL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12669806
|
Epredia
MS-143-R7 |
7 mL | Unconjugated | Prediluted | Prediluted | ||||
Beschreibung
Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Mouse
Clone: L1C1
Isotype: IgG1/k
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: B-lymphoma cells
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spezifikation
| Kappa Light Chain Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Mouse | |
| Hematopoietic | |
| B lymphoma cells | |
| Primary |
| L1C1 | |
| Monoclonal | |
| IVD | |
| Human | |
| IgG2a κ |