Learn More
Myeloperoxidase (MPO) Rabbit Polyclonal Antibody, Epredia™
Myeloperoxidase (MPO), Rabbit Polyclonal Ab is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Marke: Epredia RB-373-A0
Beschreibung
Myeloperoxidase is an important enzyme used by granulocytes during phagocytic lysis of foreign particles engulfed. In normal tissues and in a variety of myeloproliferative disorders myeloid cells of both neutrophilic and eosinophilic types, at all stages of maturation, exhibit strong cytoplasmic reactivity for MPO. Erythroid precursors, megakaryocytes, lymphoid cells, mast cells, and plasma cells are nonreactive. MPO is not observed in the neoplastic cells of a wide variety of epithelial tumors and sarcomas. MPO is useful in differentiating between myeloid and lymphoid leukemias.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified human granulocytic MPO
Positive Control: Tonsil or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spezifikation
| Myeloperoxidase (MPO) | |
| Polyclonal | |
| Unconjugated | |
| Purified human granulocytic MPO | |
| IVD | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Rabbit | |
| 100 μL | |
| Enzyme | |
| Human, Mouse | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.