missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ S100 Protein Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Epredia™ S100 Protein Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Marke:  Epredia™ RB-044-R7

Weitere Details : Netto-Gewicht : 0.00700kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 12622747

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Cow, Kangaroo, Mouse, Rat and Chicken. Others not tested.

Epitope: Not determined

Immunogen: S100 protein purified from cow brain

Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)

Positive Control: Melanoma or Schwannoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunocytochemistry

Note:

USA: RUO; Int'l: RUO

Spezifikation

Spezifikation

S100 Protein Ab-2
Polyclonal
Rabbit
7 mL
Cancer and Tumor Biology
Bovine, Chicken, Human, Kangaroo, Mouse
IgG
Immunohistochemistry (Paraffin)
Unconjugated
S100 protein purified from cow brain
RUO
Primary
Prediluted
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote